Nickel(II) perchlorate hydrate ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44316-09
44316-18
CAAA44316-09EA
105.39
CAD
CAAA44316-09
CAAA44316-18
CAAA44316-30
Nickel(II) perchlorate hydrate ≥99.998% (metals basis)
Nickel(II) perchlorate hydrate
MDL: MFCD00149804
Purity: 99.998%
CAS No.: 13637-71-3
Molecular Formula: Cl2NiO8.xH2O
Formula Weight: 257.59(anhy)
Physical Form: Crystalline
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: Green
Odor: Odorless
Melting Point: 140°
UN No.: 1481
MDL No.: MFCD00149804
Formula:
Cl₂NiO₈·xH₂O MW: 257.59 g/mol (anhy) Melting Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149804 CAS Number: 13637-71-3 EINECS: 237-124-1 UN: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...