Zinc perchlorate hexahydrate ≥99.997% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

44315-09 44315-18
CAAA44315-09EA 126.72 CAD
CAAA44315-09 CAAA44315-18 CAAA44315-30
Zinc perchlorate hexahydrate ≥99.997% (metals basis)
Zinc perchlorate hexahydrate

MDL: MFCD00150370


Purity: 99.997% CAS No.: 10025-64-6 Molecular Formula: Cl2O8Zn.6H2O Formula Weight: 372.37 (264.28anhy) Physical Form: Crystalline Density: 2.25 Refractive Index: 1.50 Appearance: White Odor: Odorless Melting Point: 106° Merck Reference: 14,10148 UN No.: 1481 MDL No.: MFCD00150370

Formula: Cl₂O₈Zn·6H₂O
MW: 372.38
Melting Pt: 106
Density: 2.25
Storage Temperature: Ambient
MDL Number: MFCD00150370
CAS Number: 10025-64-6
EINECS: 237-122-0
UN: 1481
ADR: 5.1 II
Merck Index: 14,10148

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR