Aluminium chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44313-14
44313-22
CAAA44313-14EA
368.14
CAD
CAAA44313-14
CAAA44313-22
Aluminium chloride, ultra dry ≥99.999% (metals basis)
Aluminium chloride
MDL: MFCD00003422
Fieser: 1,24 12,26 13,15 14,21 15,10 16,10 17,15 18,18 19,11 20,13
Purity: 99.999%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Powder, Ampouled under argon
Density: 2.44
pH: 2,4
Melting Point: 190°subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422
Formula:
AlCl₃ Boiling Pt: 180 °C (1013 hPa) Melting Pt: 190 °C (subl.) Density: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00003422 CAS Number: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II Merck Index: 14,337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...