Sodium periodate 99.8-100.3% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
44309-22
44309-36
44309-A3
CAAA44309-22EA
225.94
CAD
CAAA44309-22
CAAA44309-36
CAAA44309-A3
Sodium periodate 99.8-100.3% ACS
Sodium periodate
MDL: MFCD00003534
Purity: 99.8-100.3%
CAS No.: 7790-28-5
Molecular Formula: INaO4
Formula Weight: 213.89
Physical Form: Powder
Density: 3.86
Appearance: White
Odor: Odorless
Melting Point: 300° dec.
Merck Reference: 14,8640
UN No.: 1479
MDL No.: MFCD00003534
Formula:
NaIO₄ Boiling Pt: 300 °C (1013 hPa) Melting Pt: 300 °C (dec.) Density: 3.86 Storage Temperature: Ambient |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 EINECS: 232-197-6 UN: 1479 ADR: 5.1,II Merck Index: 14,8640 |
Specification Test Results
Other halogens (as Cl) | 0.02% |
Mn | 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...