3-Nitro-p-cresol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-3-nitrophenol, 1-Hydroxy-4-methyl-3-nitrobenzene

44285-09 44285-18
CAAA44285-04EA 77.1 CAD
CAAA44285-04 CAAA44285-09 CAAA44285-18 CAAA44285-30
3-Nitro-p-cresol 98%
3-Nitro-p-cresol

MDL: MFCD00007244


Purity: 98% CAS No.: 2042-14-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Physical Form: Powder Odor: Phenol-like UN No.: 2446 MDL No.: MFCD00007244

Formula: C₇H₇NO₃
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007244
CAS Number: 2042-14-0
EINECS: 218-044-6
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR