Calcium chloride, dry_ultra 99,99 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

44280-09 44280-18 44280-30
CAAA44280-09EA 364.98 CAD
CAAA44280-09 CAAA44280-18 CAAA44280-30
Calcium chloride, dry_ultra 99,99 Metals basis
Calcium chloride

MDL: MFCD00010903


Purity: 99.99% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: Powder, Ampouled under argon Density: 2.15 Melting Point: 782° Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.15
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR