Potassium hydroxide ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44273-22
44273-36
CAAA44273-22EA
159.58
CAD
CAAA44273-22
CAAA44273-36
Potassium hydroxide ≥99.98% (metals basis)
Potassium hydroxide
MDL: MFCD00003553
Notes: Contains Na less than 100ppm
Fieser: 1,935 4,409 5,557 6,486 7,303 8,415 9387 11,439 21,360
Purity: 99.98%
CAS No.: 1310-58-3
Molecular Formula: KOH
Formula Weight: 56.11
Physical Form: Pellets
Density: 2.044
Appearance: White
Odor: Odorless
pH: 13,5
Melting Point: 360°
Boiling Point: 1320-1324°
Merck Reference: 14,7640
UN No.: 1813
MDL No.: MFCD00003553
Formula:
KOH Boiling Pt: 1320…1324 °C Melting Pt: 360 °C Density: 2.044 Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8,II REACH: 01-2119487136-33 Merck Index: 14,7640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...