Tetrapotassium diphosphate, anhydrous ≥96%

Supplier: Thermo Scientific Chemicals
Warning

44269-A1 44269-A7
CAAA44269-A1EA 87.85 CAD
CAAA44269-A1 CAAA44269-A7
Tetrapotassium diphosphate, anhydrous ≥96%
tetra-Potassium diphosphate

MDL: MFCD00011393


Purity: 96% min CAS No.: 7320-34-5 Molecular Formula: K4O7P2 Formula Weight: 330.34 Physical Form: Granular Density: 2.33 Appearance: White Melting Point: 1090° Boiling Point: dec. Merck Reference: 14,7663 MDL No.: MFCD00011393

Formula: K₄O₇P₂
Boiling Pt: Dec.
Melting Pt: 1090 °C
Density: 2.33
Storage Temperature: Ambient
MDL Number: MFCD00011393
CAS Number: 7320-34-5
EINECS: 230-785-7
Merck Index: 14,7663

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR