Zinc oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44264-14 44264-22
CAAA44264-14EA 164.32 CAD
CAAA44264-14 CAAA44264-22
Zinc oxide ≥99.9% (metals basis)
Zinc oxide

MDL: MFCD00011300


Purity: 99.9% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 10-12mm dia x 4-5mm thick, Tablets, ≈2.3g/tablet Odor: Odorless Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR