Zirconium ≥99.5% (metals basis excluding Hf), foil, annealed, Thickness: 0.711 mm (0.028 in)

Supplier: Thermo Scientific Chemicals

44253-GH 44253-HP
CAAA44253-GHEA 218.04 CAD
CAAA44253-GH CAAA44253-HP
Zirconium ≥99.5% (metals basis excluding Hf), foil, annealed, Thickness: 0.711 mm (0.028 in)
Zirconium

MDL: MFCD00011303


Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈46.2g/100x100mm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR