Zinc bromide, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44223-09
44223-18
CAAA44223-09EA
140.94
CAD
CAAA44223-09
CAAA44223-18
CAAA44223-30
Zinc bromide, anhydrous ≥99.999% (metals basis)
Zinc bromide
MDL: MFCD00011294
Fieser: 2,463 13,349 15,368 16,389 18,409 19,409 20,438 21,493
Purity: 99.999%
CAS No.: 7699-45-8
Molecular Formula: ZnBr2
Formula Weight: 225.19
Physical Form: Lumps, Packaged under argon
Density: 4.201
Refractive Index: 1.5452
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 394°
Boiling Point: 650°
Merck Reference: 14,10129
UN No.: 3260
MDL No.: MFCD00011294
Formula:
Br₂Zn Boiling Pt: 650 °C Melting Pt: 394 °C Density: 4.201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 14,10129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...