Docusate sodium ≥96%

Supplier: Thermo Scientific Chemicals
Warning

44203-22 44203-36
CAAA44203-22EA 62.85 CAD
CAAA44203-22 CAAA44203-36
Docusate sodium ≥96%
Docusate sodium

Wetting and solubilizing agent


Purity: 96% CAS No.: 577-11-7 Molecular Formula: C20H37NaO7S Formula Weight: 444.55 Physical Form: Solid Density: 1.1 Melting Point: 153-157° Solubility: Soluble in water, acetone, alcohol Merck Reference: 14,3401 MDL No.: MFCD00012455


Wetting and solubilizing agent

Formula: C₂₀H₃₇NaO₇S
Boiling Pt: ∼180 °C (1013 hPa)
Melting Pt: 153…157 °C
Density: 1.1
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 14,3401

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR