Lithium iodide, ultra dry ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
44159-09
44159-18
44159-30
CAAA44159-09EA
99.07
CAD
CAAA44159-09
CAAA44159-18
CAAA44159-30
Lithium iodide, ultra dry ≥99% (metals basis)
Lithium iodide
MDL: MFCD00011092
Purity: 99%
CAS No.: 10377-51-2
Molecular Formula: ILi
Formula Weight: 133.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.076
Refractive Index: 1.955
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 449°
Boiling Point: 1170-1190°
Merck Reference: 14,5535
MDL No.: MFCD00011092
Formula:
ILi Boiling Pt: 1170…1190 °C Melting Pt: 449 °C Density: 4.076 Storage Temperature: Ambient |
MDL Number:
MFCD00011092 CAS Number: 10377-51-2 EINECS: 233-822-5 Merck Index: 14,5535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...