Lithium methoxide 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithium methanolate, Lithium methylate

44129-14 44129-22 44129-36
CAAA44129-14EA 114.39 CAD
CAAA44129-14 CAAA44129-22 CAAA44129-36
Lithium methoxide 98+%
Lithium methoxide

MDL: MFCD00036357 Beilstein Registry No.: 3552389


Purity: 98+% CAS No.: 865-34-9 Molecular Formula: CH3LiO Formula Weight: 37.98 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 500° dec. Beilstein Reference: 3552389 UN No.: 3206 MDL No.: MFCD00036357

Formula: CH₃LiO
MW: 37.98 g/mol
Boiling Pt: 64.6
Melting Pt: >500 °C (dec.)
Density: 0.865 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036357
CAS Number: 865-34-9
EINECS: 212-737-7
UN: 3206
ADR: 4.2 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR