ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)

Supplier: Thermo Scientific Chemicals

44126-03 44126-MD
CAAA44126-03EA 575.12 CAD
CAAA44126-03 CAAA44126-MD
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)

MDL: MFCD00036973


CAS No.: 1184-16-3 Molecular Formula: C21H27N7NaO17P3 Formula Weight: 765.40 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 4779954 MDL No.: MFCD00036973

Formula: C₂₁H₂₇N₇NaO₁₇P₃
Melting Pt: 175 to 178 °C
Storage Temperature: Freezer
MDL Number: MFCD00036973
CAS Number: 1184-16-3
EINECS: 214-664-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR