Magnesium oxide ≥96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Magnesia usta
44078-22
44078-36
44078-A1
CAAA44078-22EA
121.82
CAD
CAAA44078-22
CAAA44078-36
CAAA44078-A1
Magnesium oxide ≥96%
Magnesium oxide
MDL: MFCD00011109
Purity: 96% min
CAS No.: 1309-48-4
Molecular Formula: MgO
Formula Weight: 40.30
Physical Form: -325 Mesh Powder
Density: 3.58
Refractive Index: 1.736
Odor: Odorless
Melting Point: 2852°
Boiling Point: 3600°
Merck Reference: 14,5677
MDL No.: MFCD00011109
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 °C Melting Pt: 2852 °C Density: 3.58 Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 14,5677 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...