Bismuth(III) telluride ≥99.98% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

44077-06 44077-14 44077-22
CAAAA44077-06EA 118.97 CAD
CAAAA44077-06 CAAA44077-14 CAAA44077-22
Bismuth(III) telluride ≥99.98% (metals basis)
Bismuth (III) telluride

MDL: MFCD00014201


Purity: 99.98% CAS No.: 1304-82-1 Molecular Formula: Bi2Te3 Formula Weight: 800.76 Physical Form: Powder Appearance: Grey Odor: Odorless Melting Point: 573° UN No.: 3284 MDL No.: MFCD00014201

Formula: Bi₂Te₃
Melting Pt: 573 °C
Density: 7.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014201
CAS Number: 1304-82-1
EINECS: 215-135-2
UN: 3284
ADR: 6.1,III
Merck Index: 12,01331

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR