Formamide 99+% for spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Danger
|
44066-AP
44066-AY
CAAA44066-AEEA
72.32
CAD
CAAA44066-AE
CAAA44066-AP
CAAA44066-AY
Formamide 99+% for spectrophotometry
Formamide
MDL: MFCD00007941
Notes: Water 1 0.3%
Purity: 99+%
CAS No.: 75-12-7
Molecular Formula: CH3NO
Formula Weight: 45.04
Flash Point: 154°(310°F)
Physical Form: Liquid
Density: 1.134
Refractive Index: 1.4475
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Ammonia-like
Melting Point: 2°
Boiling Point: 210°
Beilstein Reference: 505995
Merck Reference: 14,4237
MDL No.: MFCD00007941
Formula:
HCONH₂ Boiling Pt: 210 °C Melting Pt: 2 °C Density: 1.134 Flash Pt: 154 °C (310 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 Merck Index: 14,4237 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...