N-Methyl-2-pyrrolidone (NMP) 99,5, Biograde Biochemistry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
Suitable for Biotech applications
Purity: 99.5%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid, packaged under Argon
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colorless to pale yellow
Odor: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Suitable for Biotech applications
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 202 Melting Pt: -24 Density: 1.033 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...