Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
44057-09
44057-18
CAAA44057-09EA
134.14
CAD
CAAA44057-09
CAAA44057-18
Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium (III) bromide hydrate
MDL: MFCD00049613
Purity: 99.99%
CAS No.: 15163-03-8
Molecular Formula: YbBr3.xH2O
Formula Weight: 412.77(anhy)
Physical Form: Lump
Formula:
Br₃Yb·H₂O MW: 430.77 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049613 CAS Number: 15163-03-8 EINECS: 237-350-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...