Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

44051-DJ 44051-DM 44051-HF
CAAA44051-DJEA 565.64 CAD
CAAA44051-DJ CAAA44051-DM CAAA44051-HF
Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Neodymium

MDL: MFCD00011130


Purity: 99.5% CAS No.: 7440-00-8 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.22g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Nd
MW: 144.24
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR