Holmium ≥99.9% (metals basis), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

44050-DJ 44050-HF
CAAA44050-DJEA 1482.04 CAD
CAAA44050-DJ CAAA44050-HF
Holmium ≥99.9% (metals basis), rods, Ø 6.35 mm (0.25 in)
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.28g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR