Cerium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals
Danger

44037-DJ 44037-DM 44037-HF
CAAA44037-DJEA 668.34 CAD
CAAA44037-DJ CAAA44037-DM CAAA44037-HF
Cerium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Cerium

MDL: MFCD00010924

Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3468
Melting Pt: 795
Density: ∼6.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR