Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gadolinium (III) oxide

44020-14 44020-22 44020-36
CAAA44020-14EA 235.42 CAD
CAAA44020-14 CAAA44020-22 CAAA44020-36
Digadolinium trioxide ≥99.99% (REO, rare earth oxide basis), nanopowder
Digadolinium trioxide

MDL: MFCD00011026


Purity: 99.99+% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 20-40nm Powder Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026

Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2330 °C
Density: 7.407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Merck Index: 14,4326

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR