Aluminium oxide, regular desiccant

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

44018-22 44018-36 44018-A3
CAAA44018-22EA 54.98 CAD
CAAA44018-22 CAAA44018-36 CAAA44018-A3
Aluminium oxide, regular desiccant
Aluminium oxide

Dry gases and liquids. Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2. Liquids include: benzene, hexane, oil, pentane.


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 3.2mm Spheres Odor: Odorless MDL No.: MFCD00003424


Dry gases and liquids. Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2. Liquids include: benzene, hexane, oil, pentane.

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR