Lead ≥99.9% (metals basis), wire, Ø 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44016-G6
44016-H2
CAAA44016-G6EA
176.96
CAD
CAAA44016-G6
CAAA44016-H2
Lead ≥99.9% (metals basis), wire, Ø 2.0 mm (0.08 in)
Lead
MDL: MFCD00134050
Purity: 99.9%
CAS No.: 7439-92-1
Physical Form: ≈35.6g/m
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00134050
Formula:
Pb Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Specification Test Results
Form | ~ 35.6g/m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...