Copper ≥99.9% (metals basis), turnings

Supplier: Thermo Scientific Chemicals

44001-18 44001-30 44001-A1
CAAA44001-18EA 80.74 CAD
CAAA44001-18 CAAA44001-30 CAAA44001-A1
Copper ≥99.9% (metals basis), turnings
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: Typically 1-12mm wide Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR