Diethyl phosphonate 97+%

Supplier: Thermo Scientific Chemicals
Danger

44000-14 44000-22 44000-36 44000-A4
CAAA44000-14EA 71.73 CAD
CAAA44000-14 CAAA44000-22 CAAA44000-36 CAAA44000-A4
Diethyl phosphonate 97+%
Diethyl phosphonate

MDL: MFCD00044573


Purity: 97+% CAS No.: 762-04-9 Molecular Formula: (C2H5O)2P(O)H Formula Weight: 138.10 Flash Point: 74°(165°F) Physical Form: Liquid, Packaged under argon Density: 1.070 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Weak Melting Point: -70° Boiling Point: 50-51°/2mm MDL No.: MFCD00044573

Formula: C₄H₁₁O₃P
Boiling Pt: 50…51 °C (2 mmHg)
Melting Pt: –70 °C
Density: 1,070
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044573
CAS Number: 762-04-9
EINECS: 212-091-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR