Dimethyl sulfoxide, anhydrous ≥99.8%, ChemSeal™
Supplier: Thermo Scientific Chemicals
Warning
|
Packaged under argon in resealable ChemSeal™ bottles.
43998-K2
43998-M1
CAAA43998-5YEA
1360.05
CAD
CAAA43998-5Y
CAAA43998-AE
CAAA43998-K2
CAAAA43998-M1
Dimethyl sulfoxide, anhydrous ≥99.8%, ChemSeal™
Dimethyl sulphoxide
Anhydrous
Purity: 99.8+%
CAS No.: 67-68-5
Molecular Formula: (CH3)2SO
Formula Weight: 78.13
Flash Point: 87°(189°F)
Physical Form: Liquid
Density: 1.101
Refractive Index: 1.4790
Sensitivity: Hygroscopic
Odor: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formula:
(CH₃)₂SO Boiling Pt: 189 °C Melting Pt: 18.4 °C Density: 1.101 Flash Pt: 87 °C (189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |
|
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...