Tungsten ≥99.95% (metals basis), wire, annealed, Ø 0.375 mm (0.015 in)

Supplier: Thermo Scientific Chemicals

43995-G5 43995-H1
CAAA43995-G5EA 105.7 CAD
CAAA43995-G5 CAAA43995-H1
Tungsten ≥99.95% (metals basis), wire, annealed, Ø 0.375 mm (0.015 in)
Tungsten

MDL: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR