Potassium tetrachloroplatinate(II) ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate
43946-03
43946-06
CAAA43946-03EA
281.24
CAD
CAAA43946-03
CAAA43946-06
Potassium tetrachloroplatinate(II) ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®
Potassium tetrachloroplatinate (II)
MDL: MFCD00011378
Purity: 99.99%
CAS No.: 10025-99-7
Molecular Formula: Cl4K2Pt
Formula Weight: 415.11
Physical Form: Crystalline
Density: 3.38
Appearance: Red-brown
Odor: Odorless
Melting Point: dec.
Merck Reference: 14,7682
UN No.: 3288
MDL No.: MFCD00011378
Formula:
Cl₄K₂Pt MW: 415.09 g/mol Boiling Pt: 35…40 °C (20 mmHg) Melting Pt: Dec. Density: 3.38 Storage Temperature: Ambient |
MDL Number:
MFCD00011378 CAS Number: 10025-99-7 EINECS: 233-050-9 UN: 3288 ADR: 6.1,III Merck Index: 14,7682 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...