Phenyl propargyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43923-03
43923-06
43923-14
CAAA43923-03EA
40.45
CAD
CAAA43923-03
CAAA43923-06
CAAA43923-14
CAAA43923-22
Phenyl propargyl ether 97%
Phenyl propargyl ether
MDL: MFCD00048107
Purity: 97%
CAS No.: 13610-02-1
Molecular Formula: C9H8O
Formula Weight: 132.16
Physical Form: Liquid
Density: 1.030
Refractive Index: 1.5330
Appearance: Colorless to yellow
Odor: Pungent
Boiling Point: 89-90°/14mm
MDL No.: MFCD00048107
Formula:
C₉H₈O Boiling Pt: 89…90 °C (14 mmHg) Density: 1,030 Storage Temperature: Ambient |
MDL Number:
MFCD00048107 CAS Number: 13610-02-1 EINECS: 237-095-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...