1-tert-Butyl-4-ethynylbenzene 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-Butyl)-4-ethynylbenzene, (4-tert-Butylphenyl)ethyne, (4-tert-Butylphenyl)acetylene, 4-tert-Butylphenylacetylene, 1-Ethynyl-4-tert-butylbenzene

43920-04 43920-09
CAAA43920-04EA 60.36 CAD
CAAA43920-04 CAAA43920-09
1-tert-Butyl-4-ethynylbenzene 90+%
1-tert-Butyl-4-ethynylbenzene

MDL: MFCD00190197


Purity: 90+% CAS No.: 772-38-3 Molecular Formula: C12H14 Formula Weight: 158.24 Flash Point: 82°(180°F) Physical Form: Liquid Density: 0.878 Refractive Index: 1.5297 Odor: Characteristic Boiling Point: 130°/40mm UN No.: 3082 MDL No.: MFCD00190197

Formula: C₁₂H₁₄
MW: 158.24 g/mol
Boiling Pt: 130 °C (40 mmHg)
Density: 0,878
Flash Pt: 82 °C (180 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190197
CAS Number: 772-38-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR