Ammonium cobalt (II) sulfate hexahydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43902-22
43902-36
CAAA43902-22EA
230.68
CAD
CAAA43902-22
CAAA43902-36
Ammonium cobalt (II) sulfate hexahydrate 99+%
Ammonium cobalt (II) sulphate hexahydrate
As spectroscopic standard, cobalt plating, catalysis, ceramics
Purity: 99+%
CAS No.: 13586-38-4
Molecular Formula: H8CoN2O8S2.6H2O
Formula Weight: 395.22 (287.14anhy)
Physical Form: Crystalline
Density: 1.902
Appearance: Pink to red
Odor: Odorless
Melting Point: ca 120° dec.
Solubility: Soluble in water, Insoluble in alcohol
UN No.: 3077
MDL No.: MFCD00151049
As spectroscopic standard, cobalt plating, catalysis, ceramics
Formula:
H₈CoN₂O₈S₂·6H₂O Melting Pt: ∼ 120 °C (dec.) Density: 1,902 Storage Temperature: Ambient |
MDL Number:
MFCD00151049 CAS Number: 13586-38-4 EINECS: 237-043-1 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...