Ninhydrin, REacton® ACS

Supplier: Thermo Scientific Chemicals
Warning

43846-14 43846-22 43846-36
CAAA43846-14EA 115.61 CAD
CAAA43846-14 CAAA43846-22 CAAA43846-36
Ninhydrin, REacton® ACS
Ninhydrin

Reagent for determination of amino acids and amine containing compounds, as an intermediate.


CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: Pale yellow to pale brown Melting Point: 251° dec. Solubility: Soluble in water and alcohol Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791


Reagent for determination of amino acids and amine containing compounds, as an intermediate.

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 251 °C (dec.)
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 14,6554
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to brownish-white crystals
Identification and melting point Pass test.
Solubility Pass test
Sensitivity to amino acids Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR