Cerium(IV) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

43818-09 43818-18
CAAA43818-09EA 86.11 CAD
CAAA43818-09 CAAA43818-18
Cerium(IV) oxide ≥99.9% (metals basis)
Cerium (IV) oxide

for vacuum deposition


Purity: 99.9% CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 3-6mm Pieces Density: 7.132 Appearance: Pale yellow Odor: Odorless Melting Point: 2600° Merck Reference: 14,1989 MDL No.: MFCD00010933


for vacuum deposition

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.132
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 14,1989

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR