Aluminium ≥99.85% (metals basis), wire, hard, Ø 0.013 mm (0.0005 in)
Supplier: Thermo Scientific Chemicals
43811-G1
43811-G5
CAAA43811-G1EA
461.36
CAD
CAAA43811-G1
CAAA43811-G5
Aluminium ≥99.85% (metals basis), wire, hard, Ø 0.013 mm (0.0005 in)
Aluminium
MDL: MFCD00134029
Purity: 99.85%
CAS No.: 7429-90-5
Physical Form: ≈0.00036g/m, Temper: hard
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...