Magnesium silicate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
43807-18
43807-30
CAAA43807-18EA
372.88
CAD
CAAA43807-18
CAAA43807-30
Magnesium silicate ≥99% (metals basis)
Magnesium silicate
MDL: MFCD00078249
Insoluble in water
Purity: 99%
CAS No.: 1343-88-0
Molecular Formula: Mg2O4Si
Formula Weight: 140.71
Physical Form: -325 Mesh Powder
Density: 3.21
Appearance: White
Odor: Odorless
Melting Point: 1910°
Solubility: Insoluble in water
MDL No.: MFCD00078249
Formula:
MgO₃Si Melting Pt: 1910 °C Density: 3.21 Storage Temperature: Ambient |
MDL Number:
MFCD00078249 CAS Number: 1343-88-0 EINECS: 215-681-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...