Diethyl phosphonate ≥96%

Supplier: Thermo Scientific Chemicals
Danger

43801-30 43801-A1
CAAA43801-30EA 128.77 CAD
CAAA43801-30 CAAA43801-A1
Diethyl phosphonate ≥96%
Diethyl phosphonate

Intermediate for preparation of phosphonate esters


Purity: 96% CAS No.: 762-04-9 Molecular Formula: (C2H5O)2P(O)H Formula Weight: 138.10 Flash Point: 74°(165°F) Physical Form: Liquid Density: 1.070 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Weak Melting Point: -70° Boiling Point: 50-51°/2mm MDL No.: MFCD00044573


Intermediate for preparation of phosphonate esters

Formula: C₄H₁₁O₃P
Boiling Pt: 50…51 °C (2 mmHg)
Melting Pt: –70 °C
Density: 1,070
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044573
CAS Number: 762-04-9
EINECS: 212-091-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR