2-Nitro-α,α,α-trifluoro-p-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43787-04
43787-09
CAAA43787-02EA
28.91
CAD
CAAA43787-02
CAAA43787-04
CAAA43787-09
CAAA43787-18
2-Nitro-α,α,α-trifluoro-p-toluic acid 98%
2-Nitro-α,α,α-trifluoro-p-toluic acid
MDL: MFCD00007142
Purity: 98%
CAS No.: 320-94-5
Molecular Formula: C8H4F3NO4
Formula Weight: 235.12
Physical Form: Powder
Formula:
C₈H₄F₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00007142 CAS Number: 320-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...