3-Phenyl-2-propyn-1-ol 98+%
Supplier: Thermo Scientific Chemicals
43778-04
43778-09
43778-18
CAAA43778-04EA
62.09
CAD
CAAA43778-04
CAAA43778-09
CAAA43778-18
3-Phenyl-2-propyn-1-ol 98+%
3-Phenyl-2-propyn-1-ol
MDL: MFCD00040914
Beilstein Registry No.: 2040911
Purity: 98+%
CAS No.: 1504-58-1
Molecular Formula: C6H5C≡CCH2OH
Formula Weight: 132.16
Flash Point:
Formula:
C₉H₈O Boiling Pt: 129…130 °C (10 mmHg) Density: 1,078 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040914 CAS Number: 1504-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...