Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)

Supplier: Thermo Scientific Chemicals

43777-GH 43777-HP
CAAA43777-GHEA 225.94 CAD
CAAA43777-GH CAAA43777-HP
Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Aluminium

MDL: MFCD00134029


Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR