N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%, ChemSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

Packaged under argon in resealable ChemSeal™ bottles.

43741-5Y 43741-AE 43741-K2 43741-M1
CAAA43741-5YEA 303.36 CAD
CAAA43741-5Y CAAA43741-AE CAAA43741-K2 CAAAA43741-M1
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%, ChemSeal™
N-Methyl-2-pyrrolidone (NMP)

Anhydrous


Purity: 99.5% CAS No.: 872-50-4 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 86°(187°F) Physical Form: Liquid Density: 1.033 Refractive Index: 1.4705 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colorless to pale yellow Odor: Amine-like Melting Point: -24° Boiling Point: 204° Beilstein Reference: 106420 Merck Reference: 14,6117 MDL No.: MFCD00003193

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.033
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 14,6117
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR