Palladium ≥99.97%, wire, Ø 0.25 mm

Supplier: Thermo Scientific Chemicals

43738-BV 43738-BZ
CAAA43738-BVEA 94.01 CAD
CAAA43738-BV CAAA43738-BZ
Palladium ≥99.97%, wire, Ø 0.25 mm
Palladium

MDL: MFCD00011167


Purity: 99.97% CAS No.: 7440-05-3 Physical Form: ≈0.6g/m Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR