Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Supplier: Thermo Scientific Chemicals
43728-18
43728-30
43728-A1
CAAA43728-18EA
62.85
CAD
CAAA43728-18
CAAA43728-30
CAAA43728-A1
Polyvinylpyrrolidone, M.W. ∼ 1.300.000 (K85-95)
Polyvinylpyrrolidone
MDL: MFCD00149016
CAS No.: 9003-39-8
Molecular Formula: (C6H9NO)n
Physical Form: Powder
Sensitivity: Hygroscopic
Odor: Odorless
Merck Reference: 14,7697
MDL No.: MFCD00149016
Formula:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD00149016 CAS Number: 9003-39-8 Merck Index: 14,7697 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...