Palladium(II) bromide ≥99.998% (metals basis) Pd ≥ 39.5%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Palladium dibromide, Palladium bromide

43697-03 43697-06
CAAA43697-03EA 208.56 CAD
CAAA43697-03 CAAA43697-06
Palladium(II) bromide ≥99.998% (metals basis) Pd ≥ 39.5%, Premion®
Palladium (II) bromide

MDL: MFCD00011170


Purity: 99.998% CAS No.: 13444-94-5 Molecular Formula: PdBr2 Formula Weight: 266.22 Physical Form: Powder Appearance: Black Melting Point: dec. MDL No.: MFCD00011170

Formula: Br₂Pd
MW: 266.23
Melting Pt: Dec.
Density: 5.173 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011170
CAS Number: 13444-94-5
EINECS: 236-588-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR