Calcium iodide, ultra dry ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43674-14
43674-22
43674-36
CAAA43674-14EA
251.22
CAD
CAAA43674-14
CAAA43674-22
CAAA43674-36
Calcium iodide, ultra dry ≥99.99% (metals basis)
Calcium iodide
MDL: MFCD00010910
Purity: 99.99%
CAS No.: 10102-68-8
Molecular Formula: CaI2
Formula Weight: 293.89
Physical Form: Powder, Ampouled under argon
Density: 3.956
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 784°
Boiling Point: 1100°
Merck Reference: 14,1677
MDL No.: MFCD00010910
Formula:
CaI₂ Boiling Pt: 1100 °C Melting Pt: 784 °C Density: 3.956 Storage Temperature: Ambient |
MDL Number:
MFCD00010910 CAS Number: 10102-68-8 EINECS: 233-276-8 Merck Index: 14,1677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...