Copper(II) phthalocyanine
Supplier: Thermo Scientific Chemicals
43650-09
43650-18
43650-30
CAAA43650-09EA
62.57
CAD
CAAA43650-09
CAAA43650-18
CAAA43650-30
Copper(II) phthalocyanine
Copper (II) phthalocyanine
MDL: MFCD00010719
Beilstein Registry No.: 4121848
Notes: Dye content: about 95%
CAS No.: 147-14-8
Molecular Formula: C32H16CuN8
Formula Weight: 576.08
Physical Form: Powder
Appearance: Blue to purple
Melting Point: 350°
Beilstein Reference: 4121848
Merck Reference: 14,2520
MDL No.: MFCD00010719
Formula:
C₃₂H₁₆CuN₈ Melting Pt: 350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010719 CAS Number: 147-14-8 EINECS: 205-685-1 Merck Index: 14,2520 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...