Bis-trispropane 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]

43496-14 43496-22 43496-36
CAAA43496-14EA 125.29 CAD
CAAA43496-14 CAAA43496-22 CAAA43496-36
Bis-trispropane 98+%
BIS-TRIS propane

MDL: MFCD00004689 Notes: pKa1=6.8, useful pH range 6.3-9.5


Purity: 98+% CAS No.: 64431-96-5 Molecular Formula: CH2[CH2NHC(CH2OH)3]2 Formula Weight: 282.34 Physical Form: Powder Melting Point: 168-170° MDL No.: MFCD00004689

Formula: C₁₁H₂₆N₂O₆
MW: 282.34 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00004689
CAS Number: 64431-96-5
EINECS: 264-899-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR