1-(2-Bromoethyl)naphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

43491-03 43491-06
CAAA43491-03EA 143.46 CAD
CAAA43491-03 CAAA43491-06 CAAA43491-14
1-(2-Bromoethyl)naphthalene 97%
1-(2-Bromoethyl)naphthalene

MDL: MFCD00037737


Purity: 97% CAS No.: 13686-49-2 Molecular Formula: C12H11Br Formula Weight: 235.13 Flash Point: 110°(230°F) Physical Form: Liquid Boiling Point: 237-238°1.65 MDL No.: MFCD00037737

Formula: C₁₂H₁₁Br
Boiling Pt: 237…238 °C1.65
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037737
CAS Number: 13686-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR